Sodium butyrate ≥98%

Synonyms: Butyric acid sodium salt, Sodium Butyrate

303410-100G 303410-5G 303410-500G
220053-875EA 43.94 USD
220053-875 220053-877 220053-876
Sodium butyrate ≥98%
Sodium butyrate
Formula: C₄H₇NaO₂
MW: 110.09 g/mol
Melting Pt: 244 °C
Density: 1.324 g/cm³ (30 °C)
MDL Number: MFCD00002816
CAS Number: 156-54-7
EINECS: 205-857-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR