1-Chloro-3-methyl-2-butene ≥95%

303259-5G 303259-25G
220053-863EA 74.95 USD
220053-863 220053-862
1-Chloro-3-methyl-2-butene ≥95%
1-Chloro-3-methyl-2-butene
Formula: C₅H₉Cl
MW: 104.58 g/mol
Boiling Pt: 25 °C (1013 hPa)
Density: 0.9273 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000982
CAS Number: 503-60-6
EINECS: 207-972-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR