1-Butanol-[D10]

Synonyms: Propyl carbinol-D10, n-Butanol-D10, 1-Hydroxy butane-D10, 1-Butyl alcohol-D10

302996-5G 302996-1G
220053-816EA 75.38 USD
220053-816 220053-815
1-Butanol-[D10]
1-Butanol-[D10]
Formula: DO(CD₂)₃CD₃
MW: 84.04 g/mol
Boiling Pt: 116…118 °C (1013 hPa)
Melting Pt: –89 °C
Density: 0.92 g/cm³ (20 °C)
CAS Number: 34193-38-9
UN: 1120
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR