5-Methylthiazole

302740-1G 302740-5G
220053-491EA 31.23 USD
220053-491 220053-492
5-Methylthiazole
5-Methylthiazole
Formula: C₄H₅NS
MW: 99.16 g/mol
Boiling Pt: 139 °C (1013 hPa)
Melting Pt: –40.4 °C
Density: 1.127 g/cm³ (20 °C)
MDL Number: MFCD00005335
CAS Number: 3581-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR