4-Pentyn-1-ol ≥97%

Warning

Synonyms: 1-Pentyn-5-ol

302481-5G 302481-25G
220053-801EA 95.85 USD
220053-801 220053-800
4-Pentyn-1-ol ≥97%
4-Pentyn-1-ol
Formula: C₅H₈O
MW: 84.12 g/mol
Boiling Pt: 50 °C (1013 hPa)
Density: 0.91 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002974
CAS Number: 5390-04-5
EINECS: 226-383-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR