2,2-Bis(bromomethyl)-1,3-propanediol ≥98%

Synonyms: Dibromoneopentyl glycol, 2,2-Bis(bromomethyl)propane-1,3-diol

301930-1KG 301930-25G 301930-3KG
220051-168EA 156.92 USD
220051-168 220051-169 220053-771
2,2-Bis(bromomethyl)-1,3-propanediol ≥98%
2,2-Bis(bromomethyl)-1,3-propanediol
Formula: C₅H₁₀Br₂O₂
MW: 261.94 g/mol
Boiling Pt: 371 °C (1013 hPa)
Melting Pt: 107…109 °C
Density: 2.2 g/cm³ (20 °C)
MDL Number: MFCD00004688
CAS Number: 3296-90-0
EINECS: 221-967-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR