1-Chloroethyl chloroformate ≥98%

301485-25G 301485-5G
220053-737EA 113.08 USD
220053-737 220053-738
1-Chloroethyl chloroformate ≥98%
1-Chloroethyl chloroformate
Formula: C₃H₄Cl₂O₂
Boiling Pt: 118…119 °C (1013 hPa)
Melting Pt: –65 °C
Density: 1.31 g/cm³ (20 °C)
Flash Pt: 40 °C
MDL Number: MFCD00000647
CAS Number: 50893-53-3
EINECS: 256-834-2
UN: 2742
ADR: 6.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR