3-(Trifluoromethoxy)benzoic acid ≥97%

Warning

301388-5G 301388-1G
220053-729EA 65.05 USD
220053-729 220053-728
3-(Trifluoromethoxy)benzoic acid ≥97%
3-(Trifluoromethoxy)benzoic acid
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 91…92 °C
MDL Number: MFCD00041501
CAS Number: 1014-81-9
EINECS: 213-802-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR