2,6-Naphthalenedicarboxylic acid

Synonyms: 2,6-Naphthalenedicarboxylic Acid, Naphthalene-2,6-dicarboxylic acid

301353-5G 301353-25G
220053-726EA 68.82 USD
220053-726 220053-725
2,6-Naphthalenedicarboxylic acid
2,6-Naphthalenedicarboxylic acid
Formula: C₁₂H₈O₄
MW: 216.19 g/mol
Boiling Pt: 422 °C (1013 hPa)
Melting Pt: 366…368 °C
MDL Number: MFCD00004105
CAS Number: 1141-38-4
EINECS: 214-527-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR