3-Chloro-3-methyl-1-butyne ≥97%

301345-5G 301345-25G
220053-724EA 138.92 USD
220053-724 220053-723
3-Chloro-3-methyl-1-butyne ≥97%
3-Chloro-3-methyl-1-butyne
Formula: C₅H₇Cl
MW: 102.56 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00190221
CAS Number: 1111-97-3
EINECS: 214-188-9
UN: 1993
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR