D-(-)-Cysteine ≥99%

Synonyms: (S)-2-Amino-3-mercaptopropanoic acid, H-D-Cys-OH, (S)-(-)-Cysteine

30095-100MG 30095-1G
220053-592EA 83.25 USD
220053-592 220053-593
D-(-)-Cysteine ≥99%
D-(-)-Cysteine
Formula: C₃H₇NO₂S
MW: 121.16 g/mol
MDL Number: MFCD00066461
CAS Number: 921-01-7
EINECS: 213-062-0

Order Now

Specification Test Results

Assay 99% (RT)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR