Methyl chlorodifluoroacetate ≥99%

300837-25G 300837-5G
220053-587EA 89.06 USD
220053-587 220053-588
Methyl chlorodifluoroacetate ≥99%
Methyl chlorodifluoroacetate
Formula: C₃H₃ClF₂O₂
Boiling Pt: 77.5…78.5 °C (1013 hPa)
Density: 1.3701 g/cm³ (18 °C)
MDL Number: MFCD00000775
CAS Number: 1514-87-0
EINECS: 216-154-9
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR