(2S,3S)-(+)-2,3-Butanediol ≥97%

Synonyms: (2S,3S)-Butane-2,3-diol, (S,S)-(+)-2,3-Butanediol, (2S,3S)-(+)-2,3-Butylene glykol, (S,S)-(+)-2,3-Dihydroxy butane, (2S,3S)-(+)-2,3-Dihydroxy butane, (S,S)-(+)-2,3-Butylene glykol, L(+)-2,3-Butanediol

300349-1G 300349-500MG 300349-5G
220053-551EA 119.54 USD
220053-551 220053-552 220053-553
(2S,3S)-(+)-2,3-Butanediol ≥97%
(2S,3S)-(+)-2,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 179 °C (1013 hPa)
Density: 0.99 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00063648
CAS Number: 19132-06-0

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR