Methylcyclohexane, anhydrous ≥99%

Danger

300306-2L 300306-100ML 300306-1L
220053-545EA 112.31 USD
220053-545 220053-543 220053-544
Methylcyclohexane, anhydrous ≥99%
Methylcyclohexane
Formula: C₆H₁₁CH₃
MW: 98.19 g/mol
Boiling Pt: 101 °C (1013 hPa)
Melting Pt: -126 °C
Density: 0.7693 g/cm³ (20 °C)
Flash Pt: -4 °C
Storage Temperature: Ambient
MDL Number: MFCD00001497
CAS Number: 108-87-2
EINECS: 203-624-3
UN: 2296
ADR: 3,II
Merck Index: 13,06074
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f7f0d5a1c32138da16df4781897a9c0184e42b7&vid=39a9392c8629d0e2fc79134840e6d8a8dc8a76e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR