Palladium(II) trifluoroacetate ≥97%

Synonyms: Palladium(II) 2,2,2-trifluoroacetate

299685-5G 299685-1G
220053-465EA 809.85 USD
220053-465 220053-464
Palladium(II) trifluoroacetate ≥97%
Palladium(II) trifluoroacetate
Formula: C₄F₆O₄Pd
MW: 332.45 g/mol
MDL Number: MFCD00013204
CAS Number: 42196-31-6

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR