2-Acetyl-5-methylfuran

Synonyms: 1-(5-Methylfuran-2-yl)ethanone

299553-25G 299553-5G
220053-142EA 71.51 USD
220053-142 220053-143
2-Acetyl-5-methylfuran
2-Acetyl-5-methylfuran
Formula: C₇H₈O₂
MW: 124.14 g/mol
MDL Number: MFCD00003243
CAS Number: 1193-79-9
EINECS: 214-779-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR