Trifluoroacetic acid ≥99%

Danger

299537-100G 299537-500G 299537-25G
220053-139EA 64 USD
220053-139 220053-141 220053-140
Trifluoroacetic acid ≥99%
Trifluoroacetic acid
Formula: C₂HF₃O₂
Boiling Pt: 71.8 °C (1013 hPa)
Melting Pt: –15 °C
Density: 1.5351 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR