Methyl decanoate ≥99%

Synonyms: Decanoic acid methyl ester, Capric acid methyl ester, Decylic acid methyl ester, Methyl caprate

299030-100G 299030-2.5G
220051-659EA 85.18 USD
220051-659 220051-660
Methyl decanoate ≥99%
Methyl decanoate
Formula: C₁₁H₂₂O₂
MW: 186.29 g/mol
Boiling Pt: 224 °C (1013 hPa)
Melting Pt: -18 °C
Density: 0.8724 g/cm³ (20 °C)
Flash Pt: 94 °C
MDL Number: MFCD00009580
CAS Number: 110-42-9
EINECS: 203-766-6

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR