DL-Isoleucine ≥99%

Synonyms: H-DL-Ile-OH

298689-50G 298689-10G
220053-429EA 168 USD
220053-429 220053-428
DL-Isoleucine ≥99%
DL-Isoleucine
Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Melting Pt: 275 °C
Density: 1.255 g/cm³ (-123.15 °C)
MDL Number: MFCD00004268
CAS Number: 443-79-8
EINECS: 207-139-8
Merck Index: 13,05198

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR