D-(-)-Leucinol ≥98%

Synonyms: D-Leucinol, (R)-(-)-Leucinol, (R)-(-)-2-Amino-4-methylpentan-1-ol, (R)-2-Amino-4-methylpentan-1-ol

298417-1G 298417-5G
220053-420EA 40.92 USD
220053-420 220053-421
D-(-)-Leucinol ≥98%
D-(-)-Leucinol
Formula: C₆H₁₅NO
MW: 117.19 g/mol
Boiling Pt: 199 °C (1013 hPa)
Density: 0.897 g/cm³ (20 °C)
MDL Number: MFCD00004734
CAS Number: 53448-09-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR