Lithium chloride hydrate ≥99.99%

Warning

298328-25G 298328-100G
220053-099EA 224.62 USD
220053-099 220053-098
Lithium chloride hydrate ≥99.99%
Lithium chloride hydrate
Formula: ClLi·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00149764
CAS Number: 85144-11-2
EINECS: 231-212-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR