Copper(I) thiocyanate

Warning

298212-250G 298212-50G
220053-088EA 613.85 USD
220053-088 220053-089
Copper(I) thiocyanate
Copper(I) thiocyanate
Formula: CCuNS
Density: 2.846 g/cm³ (15 °C)
MDL Number: MFCD00010980
CAS Number: 1111-67-7
EINECS: 214-183-1
UN: 3077
ADR: 9,III
Merck Index: 13,02699

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR