Erbium trinitrate pentahydrate ≥99.9%

Synonyms: Erbium(III) nitrate hydrate, Erbium (III) nitrate pentahydrate

298166-100G 298166-25G
220053-081EA 481.54 USD
220053-081 220053-082
Erbium trinitrate pentahydrate ≥99.9%
Erbium trinitrate pentahydrate
Formula: ErN₃O₉·5H₂O
MW: 443.35 g/mol
MDL Number: MFCD00149691
CAS Number: 10031-51-3
EINECS: 233-436-7
UN: 1477
ADR: 5.1,III

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR