Dysprosium trinitrate hydrate ≥99.9%

Synonyms: Dysprosium (III) nitrate hydrate

298158-100G 298158-25G
220053-079EA 343.08 USD
220053-079 220053-080
Dysprosium trinitrate hydrate ≥99.9%
Dysprosium trinitrate hydrate
Formula: DyN₃O₉·H₂O
MW: 366.53 g/mol
CAS Number: 100641-13-2
EINECS: 233-410-5
UN: 1477
ADR: 5.1,II

Order Now

Specification Test Results

Assay 99.9% trace metals basis

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR