4-Bromo-3-(trifluoromethyl)aniline ≥98%

Synonyms: 5-Amino-2-bromobenzotrifluoride, 4-Bromo-α,α,α-trifluoro-m-toluidine, 5-Amino-2-bromo-α,α,α-trifluorotoluene

297712-5G 297712-25G
220053-384EA 53.85 USD
220053-384 220053-383
4-Bromo-3-(trifluoromethyl)aniline ≥98%
4-Bromo-3-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 81 - 84 °C (0.5 -rr)
Melting Pt: 55 - 56 °C
MDL Number: MFCD00007827
CAS Number: 393-36-2
EINECS: 206-885-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR