2-Amino-3-nitrophenol ≥98%

Warning

Synonyms: 2-Hydroxy-6-nitroaniline

297003-5G 297003-25G
220053-367EA 43.08 USD
220053-367 220053-366
2-Amino-3-nitrophenol ≥98%
2-Amino-3-nitrophenol
Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
MDL Number: MFCD00010875
CAS Number: 603-85-0
EINECS: 210-060-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR