n-Octane, anhydrous ≥99%
|
Danger
|
296988-20L-P2
296988-2L
296988-4X2L
296988-100ML
296988-1L
220053-360EA
726.15
USD
220053-360
220053-361
220053-362
220053-358
220053-359
n-Octane, anhydrous ≥99%
n-Octane
Formula:
H₃C(CH₂)₆CH₃ MW: 114.23 g/mol Boiling Pt: 126 °C (1013 hPa) Melting Pt: –57 °C Density: 0.703 g/cm³ (20 °C) Flash Pt: 8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009556 CAS Number: 111-65-9 EINECS: 203-892-1 UN: 1262 ADR: 3,II Merck Index: 13,06782 |
Specification Test Results
Assay | 99% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...