Ethylbenzene, anhydrous ≥99.8%

Danger

Synonyms: Phenylethane

296848-1L 296848-100ML 296848-2L
220053-345EA 27.91 USD
220053-345 220053-344 220053-346
Ethylbenzene, anhydrous ≥99.8%
Ethylbenzene
Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 136.2 °C (1013 hPa)
Melting Pt: –95 °C
Density: 0.867 g/cm³ (20 °C)
Flash Pt: 15 °C
Storage Temperature: Ambient
MDL Number: MFCD00011647
CAS Number: 100-41-4
EINECS: 202-849-4
UN: 1175
ADR: 3,II
Merck Index: 13,03801
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c67d12bb66494ff09657a029fa1e68b09af34358&vid=3e5678dd6a0342c5ec35b93e48587e8125977cf7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.8%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR