(R)-(-)-sec-Butylamine ≥99%

Danger

Synonyms: (R)-(-)-2-Butylamine, (R)-(-)-2-Aminobutane

296643-250MG 296643-1G
220053-318EA 96.28 USD
220053-318 220052-254
(R)-(-)-sec-Butylamine ≥99%
(R)-(-)-sec-Butylamine
Formula: C₄H₁₁N
MW: 73.14 g/mol
Boiling Pt: 63 °C (1013 hPa)
Melting Pt: -104 °C
Density: 0.725 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064416
CAS Number: 13250-12-9
EINECS: 236-232-6
UN: 2733
ADR: 3,II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR