Anisole, anhydrous ≥99.7%

Danger

Synonyms: Methyl phenyl ether, Methoxybenzene

296295-100ML 296295-2L 296295-1L
220053-290EA 36 USD
220053-290 220053-292 220053-291
Anisole, anhydrous ≥99.7%
Anisole
Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 154 °C (1013 hPa)
Melting Pt: –37 °C
Density: 0.9961 g/cm³ (20 °C)
Flash Pt: 44 °C
Storage Temperature: Ambient
MDL Number: MFCD00008354
CAS Number: 100-66-3
EINECS: 202-876-1
UN: 2222
ADR: 3,III
Merck Index: 13,00672
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d32206d4bb9dbd12360d11d39b31237d54a8dcf8&vid=286a07a5e34a1ec55ff6339785af95d9a4326825&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.7%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR