(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

Synonyms: (R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, (R)-(+)-BINAP, (R)-BINAP, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthalene, (R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl

295817-1G 295817-250MG 295817-5G
220053-051EA 93.15 USD
220053-051 220053-052 220053-053
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
(R)-(+)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl
Formula: C₄₄H₃₂P₂
MW: 622.69 g/mol
Melting Pt: 239…242 °C
MDL Number: MFCD00010805
CAS Number: 76189-55-4
Merck Index: 13,01223

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR