Propene ≥99%

Danger

Synonyms: Propylene

295663-100G 295663-300G
220053-023EA 285.23 USD
220053-023 220053-024
Propene ≥99%
Propene
Formula: C₃H₆
MW: 42.08 g/mol
Boiling Pt: –47.4 °C (1013 hPa)
Melting Pt: –185.2 °C
Density: 0.6513 g/cm³ (-80 °C)
Storage Temperature: Ambient
CAS Number: 115-07-1
EINECS: 204-062-1
UN: 1077
ADR: 2,
Merck Index: 12,08034

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR