2-Methylpropene ≥99%

Danger

Synonyms: 2-Methylpropene, Isobutene, Isobutylene

295469-100G 295469-1KG 295469-400G
220052-990EA 296.31 USD
220052-990 220052-991 220052-992
2-Methylpropene ≥99%
2-Methylpropene
Formula: C₄H₈
MW: 56.11 g/mol
Boiling Pt: –6.9 °C (1013 hPa)
Melting Pt: –140.3 °C
Density: 0.5942 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008898
CAS Number: 115-11-7
EINECS: 204-066-3
UN: 1055
ADR: 2,
Merck Index: 12,05155

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR