Chloroethane ≥99.7%

Danger

Synonyms: Ethyl chloride

295310-1KG 295310-500G
220052-968EA 591.92 USD
220052-968 220052-969
Chloroethane ≥99.7%
Chloroethane
Formula: C₂H₅Cl
MW: 64.51 g/mol
Boiling Pt: 12.3 °C (1013 hPa)
Melting Pt: –142 °C
Density: 0.946 g/cm³ (0 °C)
Flash Pt: –50 °C (closed cup)
Storage Temperature: Refrigerator
MDL Number: MFCD00000961
CAS Number: 75-00-3
EINECS: 200-830-5
UN: 2037
ADR: 2,
Merck Index: 12,03829

Order Now

Specification Test Results

Assay 99.7%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR