1,1-Difluoroethane ≥98%

Synonyms: FC-152a

295264-100G 295264-600G
220052-943EA 4149.23 USD
220052-943 220052-944
1,1-Difluoroethane ≥98%
1,1-Difluoroethane
Formula: C₂H₄F₂
MW: 66.05 g/mol
Boiling Pt: -25 °C (1013 hPa)
Melting Pt: -117 °C
Density: 0.6446 g/cm³ (97 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000449
CAS Number: 75-37-6
EINECS: 200-866-1
UN: 1030
ADR: 2,

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR