trans-2-Butene ≥99%

Danger

295086-200G 295086-50G
220052-904EA 2035.38 USD
220052-904 220052-905
trans-2-Butene ≥99%
trans-2-Butene
Formula: C₄H₈
MW: 56.11 g/mol
Boiling Pt: 0.88 °C (1013 hPa)
Melting Pt: –105.5 °C
Density: 0.6042 g/cm³ (20 °C)
CAS Number: 624-64-6
EINECS: 210-855-3
UN: 1012
ADR: 2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR