1-Butene ≥99%

Danger

Synonyms: 1-Butylene

295051-100G 295051-400G
220052-902EA 181.38 USD
220052-902 220052-903
1-Butene ≥99%
1-Butene
Formula: C₄H₈
MW: 56.11 g/mol
Boiling Pt: –6.3 °C (1013 hPa)
Melting Pt: –185 °C
Density: 0.6255 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00009383
CAS Number: 106-98-9
EINECS: 203-449-2
UN: 1012
ADR: 2,
Merck Index: 13,01512

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR