1,3-Butadiene

Synonyms: Buta-1,3-diene

295035-400G 295035-1KG 295035-100G
220052-901EA 440.31 USD
220052-901 220052-900 220052-899
1,3-Butadiene
1,3-Butadiene
Formula: C₄H₆
MW: 54.09 g/mol
MDL Number: MFCD00008659
CAS Number: 106-99-0
EINECS: 203-450-8
UN: 1010
ADR: 2,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR