8-Bromo-1-octanol ≥95%

Synonyms: 8-Bromooctan-1-ol, 1-Bromo-8-hydroxyoctane

294144-1G 294144-5G
220052-779EA 57.08 USD
220052-779 220052-780
8-Bromo-1-octanol ≥95%
8-Bromo-1-octanol
Formula: BrH₂C(CH₂)₆CH₂OH
MW: 209.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010388
CAS Number: 50816-19-8
EINECS: 256-785-7

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR