Trimethylsilyl trifluoromethanesulfonate ≥98.0%, Sigma-Aldrich®

Synonyms: Trimethylsilyl trifluoromethanesulphonate

29383-1KG-F 29383-250G-F
220052-083EA 175.38 USD
220052-083 220052-084
Trimethylsilyl trifluoromethanesulfonate ≥98.0%, Sigma-Aldrich®
Trimethylsilyl trifluoromethanesulphonate

Envase: produced by Wacker Chemie AG

Formula: C₄H₉F₃O₃SSi
MW: 222.26 g/mol
Boiling Pt: 77 °C (1013 hPa)
Density: 1.225 g/cm³ (20 °C)
MDL Number: MFCD00000406
CAS Number: 27607-77-8
EINECS: 248-565-4
Merck Index: 12,09849

Order Now

Specification Test Results

Assay 98.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR