2-Ethylhexyl nitrate ≥97%

293784-2.5L 293784-1L
220052-672EA 186.31 USD
220052-672 220052-671
2-Ethylhexyl nitrate ≥97%
2-Ethylhexyl nitrate
Formula: C₈H₁₇NO₃
MW: 175.23 g/mol
CAS Number: 27247-96-7
EINECS: 248-363-6
UN: 3139
ADR: 5.1,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR