Benzophenone imine ≥95%

Synonyms: Benzophenoneimine

293733-5G 293733-25G
220052-665EA 156.8 USD
220052-665 220051-798
Benzophenone imine ≥95%
Benzophenone imine
Formula: C₁₃H₁₁N
MW: 181.24 g/mol
MDL Number: MFCD00001760
CAS Number: 1013-88-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42df309ff3b8aeb0143da5777df43ebfda8afc19&vid=ae0a855ddb5d76b6aa2a87a2d032a6e5bcda309e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR