4-Chloro-3-sulfamoylbenzoic acid ≥98%

293598-1G 293598-5G
220052-658EA 66.77 USD
220052-658 220052-659
4-Chloro-3-sulfamoylbenzoic acid ≥98%
4-Chloro-3-sulphamoylbenzoic acid
Formula: C₇H₆ClNO₄S
MW: 235.65 g/mol
MDL Number: MFCD00012375
CAS Number: 1205-30-7
EINECS: 214-882-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR