Bis(2-ethylhexyl)amine ≥99%

Synonyms: Dioctylamine, Di(2-ethylhexyl)amine, Bis(2-ethylhexyl)amine

292850-1L 292850-250ML
220052-576EA 148.62 USD
220052-576 220052-577
Bis(2-ethylhexyl)amine ≥99%
Bis(2-ethylhexyl)amine
Formula: C₁₆H₃₅N
MW: 241.46 g/mol
Boiling Pt: 281.1 °C (1013 hPa)
Melting Pt: < –60 °C
Density: 0.8062 g/cm³ (20 °C)
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00009489
CAS Number: 106-20-7
EINECS: 203-372-4
UN: 2922
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR