Cyclohexene ≥99% stabilized

Danger

29240-1L 29240-500ML 29240-2.5L
220052-534EA 81.54 USD
220052-534 220052-536 220052-535
Cyclohexene ≥99% stabilized
Cyclohexene
Formula: C₆H₁₀
MW: 82.15 g/mol
Boiling Pt: 83 °C (1013 hPa)
Melting Pt: –104 °C
Density: 0.81 g/cm³ (20 °C)
Flash Pt: –20 °C
Storage Temperature: Ambient
MDL Number: MFCD00001539
CAS Number: 110-83-8
EINECS: 203-807-8
UN: 2256
ADR: 3,II
Merck Index: 13,02756

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR