2-Bromoheptane, Technical Grade

Danger

292389-100G 292389-25G
220052-532EA 322 USD
220052-532 220052-533
2-Bromoheptane, Technical Grade
2-Bromoheptane
Formula: H₃CBrCH(CH₂)₄CH₃
MW: 179.1 g/mol
CAS Number: 1974-04-5
EINECS: 217-824-3
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR