N,N'-Diphenyl-p-phenylenediamine ≥98%

Warning

Synonyms: DPPD, N,N′-Diphenyl-1,4-phenylenediamine, N,N'-Diphenyl-1,4-phenylenediamine

292265-25G 292265-5G
220052-519EA 158.23 USD
220052-519 220052-520
N,N'-Diphenyl-p-phenylenediamine ≥98%
N,N'-Diphenyl-p-phenylenediamine
Formula: C₁₈H₁₆N₂
MW: 260.34 g/mol
Boiling Pt: 469 °C (1013 hPa)
Melting Pt: 146…149 °C
Density: 1.15 g/cm³ (20 °C)
Flash Pt: 193 °C
MDL Number: MFCD00003015
CAS Number: 74-31-7
EINECS: 200-806-4
Merck Index: 12,03388

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR