1,3-Propanesultone

Danger

Synonyms: 1,2-Oxathiolane 2,2-dioxide

291250-5G 291250-25G
220052-418EA 42.32 USD
220052-418 220052-417
1,3-Propanesultone
1,3-Propanesultone
Formula: C₃H₆O₃S
MW: 122.14 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 31 °C
Density: 1.3996 g/cm³ (35 °C)
MDL Number: MFCD00005355
CAS Number: 1120-71-4
EINECS: 214-317-9
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR