Bis(2-ethylhexyl)sebacate ≥90%, Technical Grade

Synonyms: 1,10-Decanedioic acid di(2-ethylhexyl) ester, Sebacic acid di(2-ethylhexyl) ester

290831-3L 290831-25ML 290831-1L
220052-341EA 376.62 USD
220052-341 220052-340 220052-339
Bis(2-ethylhexyl)sebacate ≥90%, Technical Grade
Bis(2-ethylhexyl)sebacate
Formula: C₂₆H₅₀O₄
MW: 426.68 g/mol
Boiling Pt: 435 °C (1013 hPa)
Melting Pt: –55 °C
Density: 0.914 g/cm³ (20 °C)
Flash Pt: 213 °C
Storage Temperature: Ambient
MDL Number: MFCD00009497
CAS Number: 122-62-3
EINECS: 204-558-8
Merck Index: 13,01251
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18b3fddd17e2c2f9148bc413e5e2c4558f3bc0a9&vid=689b9837fe6dd360b0805997dc4becc04db9113e&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR