2-Ethylhexyl acrylate ≥98%

Synonyms: Acrylic acid 2-ethylhexyl ester

290815-3L 290815-18L 290815-1L 290815-25ML
220052-334EA 170 USD
220052-334 220052-331 220052-332 220052-333
2-Ethylhexyl acrylate ≥98%
2-Ethylhexyl acrylate
Formula: C₁₁H₂₀O₂
MW: 184.28 g/mol
MDL Number: MFCD00009495
CAS Number: 103-11-7
EINECS: 203-080-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR